Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Metal, 2 in. and Finer, Pieces, 99.8%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
Packaging:
Poly Bottle
7440-48-4
Co
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
[Co]
7440-48-4
Co
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
[Co]

C1292, 7440-48-4, Co

Spectrum™ Cobalt Metal, 2 in. and Finer, Pieces
CAS Max % 100%
Percent Purity 99.8%
Packaging Poly Bottle
Quantity 125 g