Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits

Manufacturer:  Alfa Aesar™ 4038722

Quantity 100g

 View more versions of this product

Catalog No. AA4038722


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

61789-51-3
MFCD02094763
UN1993
Cobalt naphthenate

Specifications

Co 6%, approximately 53% in min.eral spirits
Dark
61789-51-3
MFCD02094763
Cobalt naphthenate
40° (104°F)
100g
Liquid
UN1993

Safety and Handling

SDS & Safety Documents

GHS H Statement
H304-H351-H226-H317
May be fatal if swallowed and enters airways.
Suspected of causing cancer.
Flammable liquid and vapor.
May cause an allergic skin reaction.

GHS P Statement
P280h-P262-P301+P330+P331-P315
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : QK8925000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit