Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Nitrate, 6-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10026-22-9
CoH12N2O12
291.031
QGUAJWGNOXCYJF-UHFFFAOYSA-N
24821
CHEBI:86214
cobalt(2+);dinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Co+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 291.03
Packaging Poly Bottle
Specific Gravity 1L = 1.87kg
Quantity 500g
Chemical Name or Material Cobalt Nitrate, 6-Hydrate
Grade Baker Analyzed™ ACS Reagent