Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Nitrate, 8% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
125 mL
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 10026-22-9
Packaging Poly Bottle
Quantity 125 mL