Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10026-22-9
10026-22-9
Grade ACS
CAS Max % 1
Percent Purity 98 to 102%
Calcium (Ca) 5e-05
Chloride 2e-05
Copper (Cu) 2e-05
Insoluble Matter 0.0001
Iron (Fe) 1e-05
Lead (Pb) 2e-05
Magnesium (Mg) 5e-05
Nickel (Ni) 0.0015
Packaging Amber Glass Bottle
Potassium (K) 0.0001
Sodium (Na) 0.0005
Sulfate 5e-05
Zinc (Zn) 0.0001
Quantity 125 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.