Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt(ous) Chloride, Hexahydrate
SCI_ED

Catalog Number p-4491276
Click to view available options
Concentration or Composition (by Analyte or Components):
0.1M
1M
7791-13-1, 7732-18-5
Cl2CoH12O6
237.92
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
Cobalt (II) Chloride, Cobalt Muriate Hexahydrate
λ²-cobalt(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
7791-13-1, 7732-18-5
Cl2CoH12O6
237.92
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
Cobalt (II) Chloride, Cobalt Muriate Hexahydrate
λ²-cobalt(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

This product is for educational use - no C of A is available.

Color Pink-Purple
Quantity 500 mL
Assay Percent Range <75%w/v
Linear Formula CoCl2·6H2O
Formula Weight 237.95
Concentration or Composition (by Analyte or Components) 0.1M
Chemical Name or Material Colbalt Chloride Solution
ShelfLife 2 years
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit