Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Oxide Products for Science Education

Click to view available options
Quantity:
100g
Packaging:
Powder
1308-06-1
Co3O4
240.796
LBFUKZWYPLNNJC-UHFFFAOYSA-N
oxocobalt; oxo oxocobaltiooxy cobalt,cobalt ii oxide; cobalt iii oxide,cobalt ii,coo.co2o3,aronis24129,cobaltic tetraoxide,,cobalt ii,iii oxide, powder, <10 mum,cobalt ii oxide; oxo oxocobaltio oxy cobalt,cobalt ii,iii oxide metals basis , 2-6 micron powder,oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
11651651
oxocobalt;oxo(oxocobaltiooxy)cobalt
O=[Co].O=[Co]O[Co]=O
1308-06-1
Co3O4
240.796
LBFUKZWYPLNNJC-UHFFFAOYSA-N
oxocobalt; oxo oxocobaltiooxy cobalt,cobalt ii oxide; cobalt iii oxide,cobalt ii,coo.co2o3,aronis24129,cobaltic tetraoxide,,cobalt ii,iii oxide, powder, <10 mum,cobalt ii oxide; oxo oxocobaltio oxy cobalt,cobalt ii,iii oxide metals basis , 2-6 micron powder,oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
11651651
oxocobalt;oxo(oxocobaltiooxy)cobalt
O=[Co].O=[Co]O[Co]=O

This product is an Educational Grade Chemical — no C of A is available.

Linear Formula Co3O4
Formula Weight 240.8
Recommended Applications Education
Density 6.11g/cmsuperscript_3;
Packaging Powder
Quantity 100g
Chemical Name or Material Cobalt Oxide

ShelfLife : 5 years