Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Oxide, Black, Powder, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1308-06-1
1308-06-1
Grade Reagent
CAS Max % 1
Cobalt (Co) 0.7
Iron (Fe) 0.001
Manganese (Mn) 0.002
Nickel (Ni) 0.003
Packaging Poly Pail
Sulfate 0.005
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.