Alfa Aesar™ Cobalt oxide, typically 3.4-4.5%, Molybdenum oxide typically 11.5-14.5% on alumina

Manufacturer: Alfa Aesar™ 4557914

Quantity 25g

 View more versions of this product

Catalog No. AA4557914

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01074445
    UN Number UN3077
    Chemical Name or Material Cobalt oxide, Molybdenum oxide, On alumina

    Specifications

    Physical Form Trilobe Extrudate
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Particle Size 2.5mm
    MDL Number MFCD01074445
    UN Number UN3077
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351-H302-H319-H317
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    TSCA TSCA
    Color Blue
    Quantity 25g
    Concentration Co oxide:typically 3.4 to 4.5%; Mo oxide: typically 11.5 to 14.5%
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Cobalt oxide, Molybdenum oxide, On alumina

    GHS H Statement
    H351-H302-H319-H317
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit