Alfa Aesar™ Cobalt powder, 1.6 micron, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 1045522

Quantity 100g

 View more versions of this product

Catalog No. AA1045522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-48-4
    MDL Number MFCD00010935
    UN Number UN3089
    Chemical Name or Material Cobalt powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P284a-P261-P280-P342+P311-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Danger
    Particle Size 1.6 Micron
    Health Hazard 2 GHS H Statement
    H228-H334-H317
    Flammable solid.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Color Black
    Physical Form Powder
    MDL Number MFCD00010935
    UN Number UN3089
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    RTECS Number GF8750000
    Quantity 100g
    CAS 7440-48-4
    Odor Odorless
    Chemical Name or Material Cobalt powder
    Assay 99.8%, (metals basis)

    GHS H Statement
    H228-H334-H317
    Flammable solid.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P284a-P261-P280-P342+P311-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit