Cobalt Powder, -400 mesh, 99.95% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 44085A1

Quantity 1kg

 View more versions of this product

Catalog No. AA44085A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-48-4
    Physical Form Powder
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00010935
    Chemical Name or Material Cobalt

    Specifications

    Quantity 1kg
    Color Black
    DOT Information DOT Class: 4.1, Packaging Group: II
    EINECS Number 231-158-0
    Physical Form Powder
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00010935
    CAS 7440-48-4
    Odor Odorless
    Health Hazard 1 H228-H334-H317-H413: Flammable solid. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. May cause long lasting harmful effects to aquatic life.
    Appearance Black powder
    Chemical Name or Material Cobalt

    H228-H334-H317-H413: Flammable solid. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. May cause long lasting harmful effects to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit