Alfa Aesar™ Cobalt slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, Puratronic™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 43227QK

Quantity 25 slug

 View more versions of this product

Catalog No. AA43227QK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-48-4
    MDL Number MFCD00010935
    Chemical Name or Material Cobalt slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
    Grade Puratronic™

    Specifications

    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Danger
    Weight ≈3.6g/slug
    Physical Form Slug
    MDL Number MFCD00010935
    CAS 7440-48-4
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Quantity 25 slug
    Chemical Name or Material Cobalt slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
    Assay 99.995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit