Learn More
Microflex™ Cobalt™ X Nitrile Gloves
Extra thickness for additional protection
$36.50
Specifications
Length (English) 9.5 in.
Material Nitrile
Sterility Non-sterile
Glove Cuff Style Beaded/Rolled
Hand Ambidextrous
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-169-300
View Documents
Microflex™
N211
Small
Pack of 100 for $36.50
Case of 10 PK for $365.00
Add to cart
19-169-301
View Documents
Microflex™
N212
Medium
Pack of 100 for $36.50
Case of 10 PK for $365.00
Add to cart
19-169-302
View Documents
Microflex™
N213
Large
Pack of 100 for $36.50
Case of 10 PK for $365.00
Add to cart
19-169-303
View Documents
Microflex™
N214
X-Large
Pack of 100 for $36.50
Case of 10 PK for $365.00
Add to cart
Description
Description
  • Powder-free
  • 9.5 in. length
  • 5.5mil (palm)
  • Medical/exam grade
  • Cobalt blue color
Compliance
Meets ASTM™ D6319 standards; FDA 510(k) approved for medical use.
Specifications
Specifications
9.5 in.
Non-sterile
Ambidextrous
Nitrile
Beaded/Rolled
5.5 mil