Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COBLL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP170505
Description
COBLL1 Polyclonal specifically detects COBLL1 in Human samples. It is validated for Western Blot.Specifications
COBLL1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q53SF7 | |
COBLL1 | |
Synthetic peptides corresponding to COBLL1(COBL-like 1) The peptide sequence was selected from the middle region of COBLL1. Peptide sequence QAKPSSFFLQMQKRVSGHYVTSAAAKSVHAAPNPAPKELTNKEAERDMLP. | |
Affinity Purified | |
RUO | |
22837 | |
Human, Canine, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
COBL-like 1, COBLR1, cordon-bleu protein-like 1 | |
Rabbit | |
128 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title