Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cocarboxylase, >93% (Dry Basis), Thermo Scientific™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
154-87-0
C12H19ClN4O7P2S
460.763
MFCD00038740
YXVCLPJQTZXJLH-UHFFFAOYSA-N
thiamine pyrophosphate,cocarboxylase,thiamine pyrophosphate chloride,cocarboxilasa,cocarboxylasum,farmaneurina,pyruvodehydrase,berolase,biosyth,bioxylasi
9068
CHEBI:18290
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl phosphono hydrogen phosphate;chloride
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCOP(=O)(O)OP(=O)(O)O.[Cl-]
154-87-0
C12H19ClN4O7P2S
460.763
MFCD00038740
YXVCLPJQTZXJLH-UHFFFAOYSA-N
thiamine pyrophosphate,cocarboxylase,thiamine pyrophosphate chloride,cocarboxilasa,cocarboxylasum,farmaneurina,pyruvodehydrase,berolase,biosyth,bioxylasi
9068
CHEBI:18290
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl phosphono hydrogen phosphate;chloride
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCOP(=O)(O)OP(=O)(O)O.[Cl-]
Quantity 25g
Packaging Plastic bottle
Formula Weight 460.77
Physical Form Powder
Percent Purity ≥93%
Chemical Name or Material Cocarboxylase
Assay Dry basis