Learn More
COCO/DAND5 Mouse anti-Human, Alexa Fluor™ 594, Clone: 25D4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221979AF594
Catalog No. NP221979A94
Add to cart
Description
Description
COCO/DAND5 Monoclonal antibody specifically detects COCO/DAND5 in Human samples. It is validated for ELISA.
Specifications
Specifications
COCO/DAND5
25D4
ELISA
CER2CRL2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DANTE, DTE, FLJ38607, GREM3cerberus 2, Gremlin-3, SP1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
DAND5
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant COCO. Mouse anti human DAND5 Q8N907.
0.1 mL
Primary
199699