Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cocoa Butter, NF, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
25 kg
4 x 2.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8002-31-1
8002-31-1

CO105, 8002-31-1

Spectrum™ Cocoa Butter, NF is used as a suppository base for moisturizing the skin. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Melting Range 31-35°C
Refractive Index 1.454-1.459
Quantity 6 x 500 g
Packaging Poly Bottle
CAS Max % 100%