Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coconut Oil, Refined, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Pail
8001-31-8
8001-31-8

CO110, 8001-31-8

Spectrum™ Coconut Oil, Refined is taken from the meat of coconuts that come from the coconut palm. It is considered an edible oil and has several purposes in food, industry, and medicine. It contains high saturated fat. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Quantity 12 kg
Packaging Poly Pail
CAS Max % 100%