Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coconut Oil, Refined, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Pail
Description
Spectrum™ Chemical Coconut Oil, Refined is taken from the meat of coconuts that come from the coconut palm. It is considered an edible oil and has several purposes in food, industry, and medicine. It contains high saturated fat. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
CAS Max % | 100% |
Packaging | Drum |
Quantity | 200 L |
Grade | Reagent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction