Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cod Liver Oil, USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Drum
8001-69-2
8001-69-2

CO112, 8001-69-2

Spectrum™ Cod Liver Oil, USP is an oil from the liver of Cod fish. It is one of the best sources of omega 3 fatty acids (EPA and DHA) and contains relatively high amounts of vitamin A and vitamin D. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 4 L
Packaging Amber Glass Bottle
CAS Max % 100%