Learn More
Cod Liver Oil, USP, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Quantity:
500 mL
CO112, 8001-69-2
Spectrum™ Cod Liver Oil, USP is an oil from the liver of Cod fish. It is one of the best sources of omega 3 fatty acids (EPA and DHA) and contains relatively high amounts of vitamin A and vitamin D. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle