Learn More
Brady™ Code Reader™ 3600 (Bluetooth) Handle Configuration with Charging Station with Embedded Modem and 3 ft. USB Cable
Code Reader™ 3600 (Bluetooth) Handle Configuration with Charging Station with Embedded Modem and 3 ft. USB Cable: Code Reader™ 3600 is an ultra-fast, durable barcode reader with an aggressive scan engine. Its dual field optics make transitioning from decoding wide to small barcodes easy.
Manufacturer:  Brady™ 143561
Catalog No. 19-104-473
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Glare-reduction technology means reading barcodes on shiny surfaces is no longer a challenge
  • Features a durable quick-release, rechargeable, battery cartridge
  • By scanning the QuickConnect Code™ located on the charging station, the CR2600 establishes a Bluetooth™ connection between the reader and host device
  • The CR2600 tolerates more cleaners and disinfectants than other barcode readers on the market
Specifications
Specifications
Scanner
Safety
248.92 mm
5.1 in.
5.6 in. H x 2 in. W x 5.1 in. D
Each
5.6 in.
2 in.
Bluetooth; Charging Station; Handle; Modem; USB Cable