Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coelenterazine, native, Aequorea sp., Alfa Aesar™

Click to view available options
Quantity:
1mg
10mg
55779-48-1
C26H21N3O3
423.472
MFCD00467176
YHIPILPTUVMWQT-UHFFFAOYSA-N
coelenterazine,oplophorus luciferin,unii-3o1cb88rrd,clzn,coelenterate luciferin,coelenterazine, native,3o1cb88rrd,8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one,2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one,3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
2830
CHEBI:2311
8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O
55779-48-1
C26H21N3O3
423.472
MFCD00467176
YHIPILPTUVMWQT-UHFFFAOYSA-N
coelenterazine,oplophorus luciferin,unii-3o1cb88rrd,clzn,coelenterate luciferin,coelenterazine, native,3o1cb88rrd,8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one,2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one,3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
2830
CHEBI:2311
8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O
Packaging Plastic bottle
Quantity 1mg
Formula Weight 423.46
Physical Form Solid
Chemical Name or Material Coelenterazine