Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coenzyme A Trihydrate, Free Acid, Affymetrix/USB™

Click to view available options
Quantity:
50mg
100mg
250mg
1g
Packaging:
Plastic bottle
85-61-0
C21H36N7O16P3S
767.533
MFCD0679839
RGJOEKWQDUBAIZ-RXPJKXIYSA-N
coenzymea,coenzyme a
133639590
[[(2S,3R,4S,5S)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(3R)-3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butyl] hydrogen phosphate
CC(C)(COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)C(C(=O)NCCC(=O)NCCS)O
85-61-0
C21H36N7O16P3S
767.533
MFCD0679839
RGJOEKWQDUBAIZ-RXPJKXIYSA-N
coenzymea,coenzyme a
133639590
[[(2S,3R,4S,5S)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(3R)-3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butyl] hydrogen phosphate
CC(C)(COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)C(C(=O)NCCC(=O)NCCS)O
Quantity 250mg
Packaging Plastic bottle
Formula Weight 821.58
Physical Form Lyophilized Powder
Chemical Name or Material Coenzyme A trihydrate