Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COG2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP257681

Catalog No. NBP257681


Add to cart

Description

Description

COG2 Polyclonal antibody specifically detects COG2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

COG2
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
component of oligomeric golgi complex 2COG complex subunit 2, conserved oligomeric Golgi complex protein 2, conserved oligomeric Golgi complex subunit 2, LDLCbrefeldin A-sensitive, peripheral Golgi protein, low density lipoprotein receptor defect C complementing, Low density lipoprotein receptor defect C-complementing protein
Rabbit
IgG
100 ul
Golgi Apparatus Markers
Primary
22796
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
COG2
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VPTTASSYVDSALKPLFQLQSGHKDKLKQAIIQQWLEGTLSESTHKYYETVSDVLNSVKKMEESLKRLKQARKTTPANPVGPSGGMSDDD
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only