Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | COG5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19137420
|
Novus Biologicals
NBP19137420UL |
20 μL |
Each for $204.00
|
|
|||||
NBP191374
|
Novus Biologicals
NBP191374 |
100 μL |
Each for $482.50
|
|
|||||
Description
COG5 Polyclonal specifically detects COG5 in Human samples. It is validated for Western Blot.Specifications
COG5 | |
Polyclonal | |
Rabbit | |
Human | |
component of oligomeric golgi complex 5, FLJ44289 | |
COG5 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
NP_006339 | |
10466 | |
Synthetic peptide directed towards the N terminal of human COG5. Peptide sequence MGWVGGRRRDSASPPGRSRSAADDINPAPANMEGGGGSVAVAGLGARGSG. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title