Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Cohn Fraction V Bovine Albumin, pH 5.2

Lyophilized powder

Manufacturer:  Affymetrix/USB™ J1085222

 View more versions of this product

Catalog No. AAJ1085222

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Source: Bovine blood
  • Form: Lyophilized powder
  • Assay (electrophoresis): ≥96%
  • Protein: ≥95%
  • pH (1%, H2O): 5.0 - 5.4
  • Residue on Ignition: ≤1.5%
  • Loss On Drying: ≤6.0%
  • Heavy Metals (Pb): ≤50ppm
  • CAS: 9048-46-8
Specifications

Specifications

Lyophilized powder
100g
4°C, desiccated
Cohn Fraction V Bovine Albumin
5.0 - 5.4
Bovine blood
Plastic bottle
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit