Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Affymetrix/USB™ Cohn Fraction V Bovine Albumin, pH 5.2

Lyophilized powder

Specifications

Product Type Cohn Fraction V Bovine Albumin
Format Lyophilized powder
Source Bovine blood
Storage Requirements 4°C, desiccated
Packaging Plastic bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ1085218 View Documents Affymetrix/USB™
J1085218
50g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
AAJ1085222 View Documents Affymetrix/USB™
J1085222
100g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Source: Bovine blood
  • Form: Lyophilized powder
  • Assay (electrophoresis): ≥96%
  • Protein: ≥95%
  • pH (1%, H2O): 5.0 - 5.4
  • Residue on Ignition: ≤1.5%
  • Loss On Drying: ≤6.0%
  • Heavy Metals (Pb): ≤50ppm
  • CAS: 9048-46-8
Specifications

Specifications

Cohn Fraction V Bovine Albumin
Bovine blood
Plastic bottle
Lyophilized powder
4°C, desiccated
5.0 - 5.4
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit