Learn More
COL15A1 Antibody (OTI1C5), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270422V
Catalog No. NBP270422V
Add to Cart
Description
Description
COL15A1 Monoclonal specifically detects COL15A1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
COL15A1
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
COL15A1
Full length recombinant protein of human COL15A1 (NP_001846) produced in HEK293T cells.
0.1 mL
1306.0
Store at 4°C in the dark.
Western Blot
OTI1C5
Western Blot
alpha-1 polypeptide, collagen alpha-1(XV) chain, collagen, type XV, alpha 1, FLJ38566
Mouse
Affinity Purified
Primary
Human
IgG1
Recommendations
Recommendations