Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL15A1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270422AF647

Catalog No. N270422A647


Add to cart

Description

Description

COL15A1 Monoclonal antibody specifically detects COL15A1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

COL15A1
OTI1C5
Western Blot
Affinity Purified, Conjugated
COL15A1
Full length recombinant protein of human COL15A1 (NP_001846) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
alpha-1 polypeptide, collagen alpha-1(XV) chain, collagen, type XV, alpha 1, FLJ38566
Mouse
IgG1
0.1 mL
Primary
1306.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit