Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL4A3BP Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS5044R
Description
CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.Specifications
COL4A3BP | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CERT1 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
10087 |
CERT1 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9Y5P4 | |
2810404O15Rik; 9230101K08Rik; alpha 3 type IV collagen binding protein; AU016711; Ceramide transfer protein; ceramide transport protein; ceramide transporter; CERT; CERTL; COL4A3BP; collagen type IV alpha 3 binding protein; Collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; MRD34; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Stard11; StAR-related lipid transfer (START) domain containing 11; StAR-related lipid transfer protein 11; START domain-containing protein 11 | |
KLH conjugated synthetic peptide derived from human COL4A3BP. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title