Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL5A1, Mouse anti-Human, Clone: 6A7, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16318

Catalog No. 89-999-228


Add to cart

Description

Description

Specifications

Specifications

COL5A1
6A7
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
collagen, type V, alpha 1
-
Mouse
IgG1, kappa
RUO
Antibody
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Lyophilized (0.01% sodium azide).
Lyophilized
COL5A1
Purified recombinant human N alpha 1 (V) fragment.
50 μg
Store at 4°C on dry atmosphere, lyophilized antibodies are stable at least 2 years. After reconstitution with deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1289
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit