Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL6A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513534
Description
COL6A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (typesI, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
Specifications
COL6A1 | |
Polyclonal | |
Unconjugated | |
Col6a1 | |
AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; Col6a1; Col6a-1; Collagen alpha-1(VI) chain; collagen type VI alpha 1; collagen type VI alpha 1 chain; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1; OPLL; procollagen, type VI, alpha 1; RGD1565398; UCHMD1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
1291 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P12109 | |
Col6a1 | |
KLH conjugated synthetic peptide between 831-858 amino acids from the C-terminal region of human COL6A1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction