Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL8A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551280
Description
The antibody detects endogenous levels of total COL8A2 protein.
COL8A2 Polyclonal antibody specifically detects COL8A2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
COL8A2 | |
Polyclonal | |
Unconjugated | |
COL8A2 | |
AI429819; COL8A2; Collagen alpha-2(VIII) chain; collagen type VIII alpha 2; collagen VIII, alpha-2 polypeptide; collagen, type VIII, alpha 2; endothelial collagen; FECD; FECD1; PPCD; PPCD2; procollagen type VIII alpha 2; procollagen, type VIII, alpha 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1296, 329941 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P25067, P25318 | |
COL8A2 | |
Synthetic peptide of human COL8A2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction