Learn More
COL9A1 Rabbit anti-Human, Alexa Fluor 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP299960AF350
Catalog No. NB063676
Add to cart
Description
Description
COL9A1 Polyclonal antibody specifically detects COL9A1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
COL9A1
Alexa Fluor 350
50mM Sodium Borate
alpha-1 polypeptide, collagen, type IX, alpha 1
Produced in rabbits immunized with purified, recombinant COL9A1 (P20849-3; Met1-Pro328).
Affinity purified
RUO
Primary
1297
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human