Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL9A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | COL9A3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16893720
![]() |
Novus Biologicals
NBP16893720UL |
20 μL |
Each for $158.00
|
|
|||||
NBP168937
![]() |
Novus Biologicals
NBP168937 |
100 μL |
Each for $487.50
|
|
|||||
Description
COL9A3 Polyclonal specifically detects COL9A3 in Human samples. It is validated for Western Blot.Specifications
COL9A3 | |
Polyclonal | |
Rabbit | |
Q14050 | |
1299 | |
Synthetic peptides corresponding to COL9A3 (collagen, type IX, alpha 3) The peptide sequence was selected from the C terminal of COL9A3. Peptide sequence PGITGKPGVPGKEASEQRIRELCGGMISEQIAQLAAHLRKPLAPGSIGRP. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
collagen type IX proteoglycan, collagen, type IX, alpha 3, DJ885L7.4.1, EDM3collagen alpha-3(IX) chain, FLJ90759, IDDcollagen IX, alpha-3 polypeptide, MED | |
COL9A3 | |
IgG | |
63 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title