Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL9A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559075
Description
Immunogen sequence: PGVPGITGKP GVPGKEASEQ RIRELCGGMI SEQIAQLAAH LRKPLAPGSI GRP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes one of the three alpha chains of type IX collagen, the major collagen component of hyaline cartilage. Type IX collagen, a heterotrimeric molecule, is usually found in tissues containing type II collagen, a fibrillar collagen. Mutations in this gene are associated with multiple epiphyseal dysplasia type 3. [provided by RefSeq, Jan 2010].
Specifications
COL9A3 | |
Polyclonal | |
Unconjugated | |
COL9A3 | |
alpha 3 type IX collagen; AV006866; cb367; CO9A3; COL9A3; Collagen alpha-3(IX) chain; collagen IX, alpha-3 polypeptide; collagen type IX alpha 3; collagen type IX alpha 3 chain; collagen type IX proteoglycan; collagen, type IX, alpha 3; DJ885L7.4.1; EDM3; IDD; MED; procollagen, type IX, alpha 3; sb:cb367; si:ch73-162j3.1; wu:fa04f01 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1299 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14050 | |
COL9A3 | |
Recombinant protein corresponding to Human COL9A3. Recombinant protein control fragment (Product #RP-98343). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction