Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Colchicine, USP, 94-101%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
64-86-8
C22H25NO6
399.44
IAKHMKGGTNLKSZ-INIZCTEOSA-N
N-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
64-86-8
C22H25NO6
399.44
IAKHMKGGTNLKSZ-INIZCTEOSA-N
N-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1

CO115, 64-86-8, C22H25NO6

Spectrum™ Colchicine, USP is an anti-inflammatory used in the treatment of gout. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specific Rotation -240° to -250°
Quantity 25 g
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN1544
Percent Purity 94-101%