Learn More
CPC™ HFC35 Polysuflone Hose Barb Coupling Bodies
Free floating
$242.00 - $302.00
Specifications
Type In-line coupling; Free floating; EPDM seal options; Vacuum to 125 psi, 8.6 bar; Valve Spring: 316 Stainless Steel
Material Polysuflone
View More Specs
Products 3
Catalog Number Mfr. No. Type Tubing Size Price Quantity & Availability  
Catalog Number Mfr. No. Type Tubing Size Price Quantity & Availability  
05-113-150
View Documents
CPC™
HFCD17635
In-line coupling; Free floating; EPDM seal options; Vacuum to 125 psi, 8.6 bar; Valve Spring: 316 Stainless Steel 3/8 in.
Pack of 10 for $299.00
Add to cart
05-113-151
View Documents
CPC™
HFCD17835
In-line coupling; Free floating; EPDM seal options; Vacuum to 125 psi, 8.6 bar; Valve Spring: 316 Stainless Steel 1/2 in.
Pack of 10 for $302.00
Add to cart
05-113-152
View Documents
CPC™
HFC17835
In-line coupling; Free floating; EPDM seal options; Vacuum to 125 psi, 8.6 bar; Valve Spring: 316 Stainless Steel 1/2 in.
Pack of 10 for $242.00
Add to cart
Description
Description
Specifications
Specifications
In-line coupling; Free floating; EPDM seal options; Vacuum to 125 psi, 8.6 bar; Valve Spring: 316 Stainless Steel
Polysuflone