Learn More
CPC™ HFC35 Polysuflone Hose Barb Coupling Inserts
Free floating
Manufacturer:  CPC™ HFC22835
 View more versions of this product
Catalog No. 05-113-153
  Check Availability
Add to cart
Specifications
Description
  • Polysulfone couplings
  • Withstands demanding applications
  • UV resitant, able to withstand continued exposure to harmful UV rays
  • Ergonomic design
  • Operate in harsh environments in temperatures from -40°F to 280°F (-40°C to 138°C)
  • Pressure: Vacuum to 125psi (8.6 bar)
  • Large, shrouded thumb latch pad
  • Leads to high flow and exceptionally low spillage
Description & Specifications
Specifications
Polysuflone
1/2 in. hose barb non-valved in-line coupling insert