Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Masterflex™ Industrial Correlated Aluminum Flowmeter

Provides liquid or gas flow readings at capacity levels to 0.48 gpm of water while maintaining an accuracy of ±5% of full scale.

Supplier:  Masterflex™ MFLX0326640

Catalog No. 15-078-172

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

 

  • 65mm correlated/calibrated
  • Without valve
  • Ranges: Air, 5cc to 58L/min.; water, 0.025cc to 1.8L/min.
  • The 65mm borosilicate glass tube has fused-on ceramic scale and is enclosed in a unitized holder
  • Precision taper of tube and integral float guides ensure float fidelity
  • One-piece aluminum frame assures maximum meter protection and safety in industrial applications
  • With tungsten carbide float, front shield with 1.5X magnification for more accurate readings
  • Float/scale correlation symbol and reading edge instructions are screened on the scale window; includes calibration table for air and water. 

Certifications

Certificate indicating instrument traceability to standards provided by NIST available on request. Call your Fisher Scientific™ Customer Service Representative for details.

Specifications

Specifications

Flowmeter
6082 to 58,500 mL/min.
200 psig
1/8 in. NPT (F)
0.25% of scale reading
±5% (Full Scale)
Aluminum
NIST traceable; Certificate indicating instrument traceability to standards provided by NIST™ available on request.
176 to 1866 mL/min.
121°C
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit