Learn More
Masterflex Variable-Area Flowmeter, Correlated Reading, Dual-Scale, Polycarbonate Shield
Don’t sacrifice suitability for precision
$810.50
Specifications
Temperature Rating | 65°C |
---|
Includes: Calibration table for air and water and comes with glass and stainless-steel floats.
Catalog Number | Accuracy | Pressure Rating | Flow Rate Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Accuracy | Pressure Rating | Flow Rate Range | Price | Quantity & Availability | ||||
15-078-112
|
±2% of the reading or ±1 scale division | 100 psig | Glass: 0.2 to 36; 316 Stainless: 0.4 to 77mL/min. |
Each for $810.50
|
|
Description
Accurate to ±2% of the reading or ±1 scale division1. Repeatability: ±1% of reading or ±0.5 scale division, whichever is greater. Max. Temperature: 65°C (150°F) at full pressure rating.
For micro flowmeter, accuracy is ±5% of reading or ±2 scale division for air and ±10% of reading or ±3 scale division for water.
Specifications
65°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.