Learn More
Cole Parmer Gilmont™ Shielded Laboratory Direct-Reading Flowmeters
 View more versions of this product
Catalog No. 15-078-130
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
65°C
Glass and 316 Stainless Steel
13
±1% of reading or ±0.5 scale division, whichever is greater
±5% of reading or ±2mm of the scale length, whichever is greater
Accepts 3/8 in. (9.5mm) I.D. tubing
Glass: 2 to 300; 316 Stainless: 4 to 640mL/min.