Cole Parmer Masterflex™ L/S™ Easy-Load™ Pump Heads for Precision Tubing
Ideal for fast tubing changes and reduced maintenance
 View all options for this product
Catalog No. 13-310-119
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Type Pump Head
    For Use With (Equipment) L/S™ 13, 14, 16, 25, 17 and 18 tubing
    Roller Material Stainless Steel
    Speed 600rpm
    Case Material Polysulfone
    • Proprietary design and adjustable tubing retention allow for multiple tubing sizes
    • Easily change tubing without removing head from the drive
    • Chemical-resistant polysulfone housing with choice of rotor: durable stainless steel or economical cold-rolled steel
    • Each pump head for precision pump tubing accepts tubing sizes L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, and L/S 1
    • Mount up to four pump heads on a single drive, depending on drive specifications
    • Flow rates of 0.06 to 2300mL/min (1 to 600 pm), depending on pump tubing size
  • Description & Specifications
Related Products
Product Content Feedback