Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Masterflex™ P/S™ Precision Pump Tubing, PharMed™ BPT

Use this tubing for cell and tissue culture work and pharmaceutical applications. Masterflex™ L/S™ High-Performance Tubing, PharMed™ BPT resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable and formable. Good for light-sensitive samples.

$294.50 - $455.00

Specifications

Autoclavable Autoclavable
Color Opaque Beige
Resistance Properties Acids, Alkalies
Type Pump Tubing
For Use With FH15 and FH30 Peristaltic pump
View More Specs

Products 4
Catalog Number Flow Rate Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Flow Rate Diameter (Metric) Inner Price Quantity & Availability  
13-310-165
View Documents
1.8 to 1100mL/min. 4.8 mm
Pack for $337.50
Add to cart
 
13-310-166
View Documents
2.8 to 1700mL/min. 6.4 mm
Pack for $455.00
Add to cart
 
13-200-303
View Documents
3.8 to 2300 mL/min 7.9 mm
Pack for $294.50
Add to cart
 
13-200-304
View Documents
4.8 to 2900 mL/min 9.7 mm
Pack for $307.00
Add to cart
 
Description

Description

Masterflex™ L/S™ pump tubing is manufactured to extremely close tolerances that match our L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.

  • Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples.
  • Characteristics: Nontoxic; nonhemolytic; extremely low permeability; resists acids, alkalies, oxidizing agents, and animal and vegetable oils; heat-sealable, bondable, and formable. Opaque beige.
  • Certifications: FDA compliant and meets USP Class VI, EP, and NSF-51 requirements. Tubing is manufactured according to GMP.

Order Info

In pack of 7.6m (25 ft.)

Specifications

Specifications

Autoclavable
Acids, Alkalies
FH15 and FH30 Peristaltic pump
25 ft.
PharMed
Opaque Beige
Pump Tubing
CO2: 360, H2: 97, O2: 80, N2: 40
1520 cm
Sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit