Learn More
Cole Parmer Masterflex™ Standard Pump Head for Precision L/S™ tubing
Ideal for accurate and repeatable fluid transfer applications
 View more versions of this product
Catalog No. 13-310-261
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pump Head
Cold rolled Steel
2.5 x 2.75 x 3 in.
Accurate and repeatable fluid transfer applications
3.0
0 to 104°F
Polycarbonate
600rpm
2.8 to 1700mL/min.
6.4 x 7 x 7.6cm
Precision L/S™ 17 tubing
Tubing loading key, single channel mounting hardware and a 15 in. L of Silicone tubing
-20 to 40°C