Cole Parmer Transfer Tubing and Tubing Links for Manostat™ and CompuLab™ 3 Pumps
 View all options for this product
Catalog No. 13-310-483
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Diameter (English) Inner 0.19 in.
    Diameter (Metric) Inner 0.19 in.
    For Use With (Equipment) Barnant Manostat and CompuLab 3 Pumps
    For use with Barnant Manostat and CompuLab 3 Pumps.
    • Load pumping links inside pump for accurate flowrates
    • Extension tubing easily connects pumping links to pump system
  • Description & Specifications
Related Products
Product Content Feedback