Learn More
Cole-Parmer™ Tygon™ Lab Tubing
Use this tubing for general laboratory applications, analytical instruments and transfer of viscous fluids. Cole-Parmer™ Tygon™ Lab Tubing is environmentally responsible, more sustainable, and safer for products, people and the planet.
 View more versions of this product
Catalog No. 13-310-271
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • May be used with virtually all inorganic chemicals
  • Nonaging, nonoxidizing
  • Smooth bore
  • Low gas permeability
  • Transparent, firm material
  • Temperature range -58 to +165°F (-50 to +74°C)
Description & Specifications
Specifications
Tygon
3.1mm
16
-50° to +74°C
Yes
3.2mm
50 ft.