Learn More
Invitrogen™ COLEC11 Recombinant Rabbit Monoclonal Antibody (148)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA531088
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse CL-K1/COLEC11.
This gene encodes a member of the collectin family of C-type lectins that possess collagen-like sequences and carbohydrate recognition domains. Collectins are secreted proteins that play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. The encoded protein binds to multiple sugars with a preference for fucose and mannose. Mutations in this gene are a cause of 3MC syndrome-2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
COLEC11 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q3SXB8 | |
COLEC11 | |
Recombinant Mouse CL-K1/COLEC11 Protein (Met1-Leu272). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
148 | |
Unconjugated | |
COLEC11 | |
1010001H16Rik; 3MC2; CLK1; CL-K1; CL-K1-I; CL-K1-II; CL-K1-IIa; CL-K1-IIb; Colec11; Collectin 11; Collectin K1; collectin kidney protein 1; collectin subfamily member 11; collectin sub-family member 11; Collectin11; collectin-11; hypothetical protein LOC492459; UNQ596/PRO1182; wu:fb82e09; zgc:103572 | |
Rabbit | |
Protein A | |
RUO | |
71693 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.