Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Colistin sodium methanesulfonate

Click to view available options
Quantity:
1g
5g
8068-28-8
C58H105N16Na5O28S5
1749.811
MFCD00130824
IQWHCHZFYPIVRV-KLWRPAJRSA-I
Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate
131668505
pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16,
CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
8068-28-8
C58H105N16Na5O28S5
1749.811
MFCD00130824
IQWHCHZFYPIVRV-KLWRPAJRSA-I
Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate
131668505
pentasodium;(6S)-N-[(2R)-1-[[(2R,3S)-3-hydroxy-1-[[(2S)-1-[[(3S,6S,9S,12S,15R,18S,21S)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-8,17,20-trioxido-2,5,11,14-tetraoxo-6,9,18-tris[2-(sulfomethylamino)ethyl]-1,4,7,10,13,16,19-heptazacyclotricosa-7,16,
CCC(C)CCCCC(=NC(CCNCS(=O)(=O)O)C(=O)NC(C(C)O)C(=O)NC(CCNCS(=O)(=O)O)C(=NC1CCNC(=O)C(NC(=O)C(N=C(C(NC(=O)C(NC(=O)C(N=C(C(N=C1[O-])CCNCS(=O)(=O)O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)O)[O-])CCNCS(=O)(=O)O)C(C)O)[O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
Solubility
Freely soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Merck Index 14,2479
Solubility Information Freely soluble in water
Formula Weight 1759.89
Physical Form Powder
Color White to Yellow
Quantity 5g
Chemical Name or Material Colistin sodium methanesulfonate

RTECSNumber : GH1650000

TSCA : TSCA

storageNote1 : 2° to 8°C